missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chloropropyl)theobromine 97.0+%, TCI America™

Catalog No. C110325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C110325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C110325G Supplier TCI America Supplier No. C110325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74409-52-2
Molecular Formula C10H13ClN4O2
Molecular Weight (g/mol) 256.69
MDL Number MFCD00059926
InChI Key JMOWKXFNJSTXBW-UHFFFAOYSA-N
Synonym 1-(3-Chloropropyl)-3,7-dihydro-3,7-dimethyl-1H-purine-2,6-dione, 1-(3-Chloropropyl)-3,7-dimethylxanthine
PubChem CID 13020469
IUPAC Name 1-(3-chloropropyl)-3,7-dimethylpurine-2,6-dione
SMILES CN1C=NC2=C1C(=O)N(C(=O)N2C)CCCCl

Specifications

Melting Point 145°C
Color White-Yellow
Quantity 25 g
Formula Weight 256.69
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Chloropropyl)theobromine