missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3-Chloropropyl)piperidine Hydrochloride 98.0+%, TCI America™
$67.11 - $242.33
Chemical Identifiers
| CAS | 5472-49-1 |
|---|---|
| Molecular Formula | C8H17Cl2N |
| Molecular Weight (g/mol) | 198.131 |
| MDL Number | MFCD00012838 |
| InChI Key | OBOBUDMMFXRNDO-UHFFFAOYSA-N |
| Synonym | 1-3-chloropropyl piperidine hydrochloride, n-3-chloropropyl piperidine hydrochloride, piperidine, 1-3-chloropropyl-, hydrochloride, piperidinopropyl chloride, hydrochloride, 3-piperidinopropyl chloride hydrochloride, 1-3-chloropropyl piperidine monohydrochloride, 1-3-chloropropyl piperidine hcl, n-3-chloropropyl-piperidine hydrochloride, 1-3-chloropropyl piperidine hydrochloride 1:1, acmc-1ak9t |
| PubChem CID | 79624 |
| IUPAC Name | 1-(3-chloropropyl)piperidine;hydrochloride |
| SMILES | C1CCN(CC1)CCCCl.Cl |
Chemical Identifiers
| 5472-49-1 | |
| 198.131 | |
| OBOBUDMMFXRNDO-UHFFFAOYSA-N | |
| 79624 | |
| C1CCN(CC1)CCCCl.Cl |
| C8H17Cl2N | |
| MFCD00012838 | |
| 1-3-chloropropyl piperidine hydrochloride, n-3-chloropropyl piperidine hydrochloride, piperidine, 1-3-chloropropyl-, hydrochloride, piperidinopropyl chloride, hydrochloride, 3-piperidinopropyl chloride hydrochloride, 1-3-chloropropyl piperidine monohydrochloride, 1-3-chloropropyl piperidine hcl, n-3-chloropropyl-piperidine hydrochloride, 1-3-chloropropyl piperidine hydrochloride 1:1, acmc-1ak9t | |
| 1-(3-chloropropyl)piperidine;hydrochloride |
Specifications
| 5472-49-1 | |
| White | |
| MFCD00012838 | |
| 1-3-chloropropyl piperidine hydrochloride, n-3-chloropropyl piperidine hydrochloride, piperidine, 1-3-chloropropyl-, hydrochloride, piperidinopropyl chloride, hydrochloride, 3-piperidinopropyl chloride hydrochloride, 1-3-chloropropyl piperidine monohydrochloride, 1-3-chloropropyl piperidine hcl, n-3-chloropropyl-piperidine hydrochloride, 1-3-chloropropyl piperidine hydrochloride 1:1, acmc-1ak9t | |
| C1CCN(CC1)CCCCl.Cl | |
| 198.131 | |
| 198.13 | |
| Crystalline Powder |
| 218°C | |
| C8H17Cl2N | |
| 1 g | |
| OBOBUDMMFXRNDO-UHFFFAOYSA-N | |
| 1-(3-chloropropyl)piperidine;hydrochloride | |
| 79624 | |
| ≥98.0% (T) | |
| 1-(3-Chloropropyl)piperidine Hydrochloride |
Safety and Handling
RTECSNumber : TM6498600
TSCA : No