missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chlorophenyl)ethanol, 97%

Catalog No. AAL0478314
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0478314 25 g
AAL0478306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAL0478314 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' L0478314

CAS: 6939-95-3 | C8H9ClO | 156.609 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6939-95-3
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00021864
InChI Key QYUQVBHGBPRDKN-UHFFFAOYSA-N
Synonym 1-3-chlorophenyl ethanol, 3-chloro-alpha-methylbenzyl alcohol, 1-3-chlorophenyl ethan-1-ol, 1-3-chlorophenyl-1-ethanol, 3-chlorophenylethanol, acmc-1cap8, 1-m-chlorophenyl ethanol, 3'-chloro-1-phenylethanol, 1-3-chlorophenyl-ethanol, 1-3-chlorophenyl ethyl alcohol
PubChem CID 97779
IUPAC Name 1-(3-chlorophenyl)ethanol
SMILES CC(C1=CC(=CC=C1)Cl)O

Specifications

Density 1.173
Boiling Point 83°C to 85°C (1.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5445
Quantity 25 g
Beilstein 2042021
Formula Weight 156.61
Percent Purity 97%
Chemical Name or Material 1-(3-Chlorophenyl)ethanol
EINECSNumber 230-079-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only