missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Butenyloxy)-2,3,3,4,4,5,5-heptafluorocyclopentene 97.0+%, TCI America™

Catalog No. p-7123811
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48811G 1 g
B48815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48811G Supplier TCI America Supplier No. B48811G

Chemical Identifiers

CAS 958032-89-8
Molecular Formula C9H7F7O
Molecular Weight (g/mol) 264.143
MDL Number MFCD00084878
InChI Key LEWPTLXFUJDZPB-UHFFFAOYSA-N
Synonym BHFC
PubChem CID 59259641
IUPAC Name 1-but-3-enoxy-2,3,3,4,4,5,5-heptafluorocyclopentene
SMILES C=CCCOC1=C(C(C(C1(F)F)(F)F)(F)F)F

Specifications

Color Colorless
Quantity 1 g
Formula Weight 264.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(3-Butenyloxy)-2,3,3,4,4,5,5-heptafluorocyclopentene