missing translation for 'onlineSavingsMsg'
Learn More

1-(3-bromopropoxy)-2-nitrobenzene, 90+%, Thermo Scientific™

Chemical Identifiers

CAS 104147-69-5
Molecular Formula C9H10BrNO3
Molecular Weight (g/mol) 260.09
MDL Number MFCD00596660
InChI Key HPZBIRIHQKSGGT-UHFFFAOYSA-N
Synonym 1-3-bromopropoxy-2-nitrobenzene, 1-3-bromo-propoxy-2-nitro-benzene
PubChem CID 7172300
IUPAC Name 1-(3-bromopropoxy)-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])OCCCBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC385240050
SDS View Documents
Thermo Scientific Chemicals
385240050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

104147-69-5
260.09
HPZBIRIHQKSGGT-UHFFFAOYSA-N
7172300
C1=CC=C(C(=C1)[N+](=O)[O-])OCCCBr
C9H10BrNO3
MFCD00596660
1-3-bromopropoxy-2-nitrobenzene, 1-3-bromo-propoxy-2-nitro-benzene
1-(3-bromopropoxy)-2-nitrobenzene

Specifications

104147-69-5
100.0
150°C
90% min. (GC)
C9H10BrNO3
MFCD00596660
1-3-bromopropoxy-2-nitrobenzene, 1-3-bromo-propoxy-2-nitro-benzene
C1=CC=C(C(=C1)[N+](=O)[O-])OCCCBr
260.09
260.09
1-(3-Bromopropoxy)-2-nitrobenzene
90.0
131°C to 132°C (0.5mmHg)
Authentic
Glass bottle
NO2C6H4O(CH2)3Br
5g
HPZBIRIHQKSGGT-UHFFFAOYSA-N
1-(3-bromopropoxy)-2-nitrobenzene
7172300
90+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only