Learn More
Thermo Scientific Chemicals 1,3-Bis[tris(hydroxymethyl)methylamino]propane, 98+%
CAS: 64431-96-5 | C11H26N2O6 | 282.337 g/mol
$147.04 - $2065.41
Chemical Identifiers
| CAS | 64431-96-5 |
|---|---|
| Molecular Formula | C11H26N2O6 |
| Molecular Weight (g/mol) | 282.337 |
| MDL Number | MFCD00004689 |
| InChI Key | HHKZCCWKTZRCCL-UHFFFAOYSA-N |
| Synonym | bis-tris propane, 1,3-bis tris hydroxymethyl methylamino propane, 2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol, 1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl, 2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol, b3p |
| PubChem CID | 125132 |
| ChEBI | CHEBI:40947 |
| IUPAC Name | 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol |
| SMILES | C(CNC(CO)(CO)CO)CNC(CO)(CO)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4349614
|
Thermo Scientific Chemicals
04349614 |
25 g |
Each for $147.04
|
|
|||||
|
AA4349622
|
Thermo Scientific Chemicals
04349622 |
100 g |
Each for $481.93
|
|
|||||
|
AA4349636
|
Thermo Scientific Chemicals
04349636 |
500 g |
Each for $2,065.41
|
|
|||||
Description
1,3-Bis[tris(hydroxymethyl)methylamino]propane is used as a buffer for polymerase chain reaction and molecular biology. It is also used with hydrochloric acid buffer for stabilization of farnesyl diphosphate isolated from a strain of saccharomyces cerevisiae. It acts as a ligand and forms dinuclear hydroxo complexes with Lanthanides(III). Further, it is useful for calibration of genetically encoded pH indicators expressed in the cytosol or mitochondria. It is also employed to investigate the influence of anions with varying nucleophilic properties on the autoxidation of oxymyoglobin. In addition to this, it is used in the purification of glucose-binding protein from membranes of sulfolobus solfataricus and as a non-zwitterionic buffer, which is used in the pH range between 6.3 and 9.5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 64431-96-5 | |
| 282.337 | |
| HHKZCCWKTZRCCL-UHFFFAOYSA-N | |
| 125132 | |
| 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol |
| C11H26N2O6 | |
| MFCD00004689 | |
| bis-tris propane, 1,3-bis tris hydroxymethyl methylamino propane, 2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol, 1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl, 2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol, b3p | |
| CHEBI:40947 | |
| C(CNC(CO)(CO)CO)CNC(CO)(CO)CO |
Specifications
| 64431-96-5 | |
| C11H26N2O6 | |
| 25 g | |
| Soluble in water. | |
| C(CNC(CO)(CO)CO)CNC(CO)(CO)CO | |
| 282.337 | |
| CHEBI:40947 | |
| ≥98% | |
| 1,3-Bis[tris (hydroxymethyl)methylamino]propane |
| 168°C to 170°C | |
| MFCD00004689 | |
| bis-tris propane, 1,3-bis tris hydroxymethyl methylamino propane, 2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol, 1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl, 2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol, b3p | |
| HHKZCCWKTZRCCL-UHFFFAOYSA-N | |
| 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol | |
| 125132 | |
| 282.34 | |
| Powder |
Safety and Handling
EINECSNumber : 264-899-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only