missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,3-Bis[tris(hydroxymethyl)methylamino]propane, 98+%

Catalog No. AA4349614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4349614 25 g
AA4349622 100 g
AA4349636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4349614 Supplier Thermo Scientific Chemicals Supplier No. 04349614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 64431-96-5 | C11H26N2O6 | 282.337 g/mol

1,3-Bis[tris(hydroxymethyl)methylamino]propane is used as a buffer for polymerase chain reaction and molecular biology. It is also used with hydrochloric acid buffer for stabilization of farnesyl diphosphate isolated from a strain of saccharomyces cerevisiae. It acts as a ligand and forms dinuclear hydroxo complexes with Lanthanides(III). Further, it is useful for calibration of genetically encoded pH indicators expressed in the cytosol or mitochondria. It is also employed to investigate the influence of anions with varying nucleophilic properties on the autoxidation of oxymyoglobin. In addition to this, it is used in the purification of glucose-binding protein from membranes of sulfolobus solfataricus and as a non-zwitterionic buffer, which is used in the pH range between 6.3 and 9.5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64431-96-5
Molecular Formula C11H26N2O6
Molecular Weight (g/mol) 282.337
MDL Number MFCD00004689
InChI Key HHKZCCWKTZRCCL-UHFFFAOYSA-N
Synonym bis-tris propane, 1,3-bis tris hydroxymethyl methylamino propane, 2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol, 1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl, 2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol, b3p
PubChem CID 125132
ChEBI CHEBI:40947
IUPAC Name 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CNC(CO)(CO)CO)CNC(CO)(CO)CO

Specifications

Melting Point 168°C to 170°C
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 282.34
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 1,3-Bis[tris (hydroxymethyl)methylamino]propane
EINECSNumber 264-899-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only