missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[tris(hydroxymethyl)amino]propane, 99%

CAS: 64431-96-5 | C11H26N2O6 | 282.33 g/mol

$137.16 - $3104.24

Chemical Identifiers

CAS 64431-96-5
Molecular Formula C11H26N2O6
Molecular Weight (g/mol) 282.33
MDL Number MFCD00004689
InChI Key HHKZCCWKTZRCCL-UHFFFAOYSA-N
Synonym bis-tris propane, 1,3-bis tris hydroxymethyl methylamino propane, 2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol, 1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl, 2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol, b3p
PubChem CID 125132
ChEBI CHEBI:40947
IUPAC Name 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC202640250
SDS View Documents
Thermo Scientific Chemicals
202640250
25 g Glass bottle
Each for $137.16
Only null left
 
AC202641000
SDS View Documents
Thermo Scientific Chemicals
202641000
100 g Glass bottle
Each for $345.11
Only null left
 
AC202642500
SDS View Documents
Thermo Scientific Chemicals
202642500
250 g Glass bottle
Each for $796.06
Only null left
 
AC202640010
SDS View Documents
Thermo Scientific Chemicals
202640010
1 kg Plastic Bottle
Each for $3,104.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

64431-96-5
282.33
HHKZCCWKTZRCCL-UHFFFAOYSA-N
125132
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
C11H26N2O6
MFCD00004689
bis-tris propane, 1,3-bis tris hydroxymethyl methylamino propane, 2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol, 1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl, 2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol, b3p
CHEBI:40947
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO

Specifications

64431-96-5
100.0
White
Authentic
Glass bottle
CH2{CH2NHC(CH2OH)3]2
25 g
bis-tris propane, 1,3-bis tris hydroxymethyl methylamino propane, 2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol, 1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl, 2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol, b3p
HHKZCCWKTZRCCL-UHFFFAOYSA-N
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
125132
282.33
Crystalline Powder
98.5
164.0°C to 165.0°C
46.33 to 47.2% C, 9.82 to 10.02% N
99%
C11H26N2O6
MFCD00004689
04,III,859
Solubility in water: soluble in cold water
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
282.33
CHEBI:40947
99%
1,3-Bis[tris(hydroxymethyl)amino]propane
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 264-899-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only