missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(tosyloxy)propane 98.0+%, TCI America™

Catalog No. b18755g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18755G 5 g
B187525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18755G Supplier TCI America Supplier No. B18755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5469-66-9
Molecular Formula C17H20O6S2
Molecular Weight (g/mol) 384.46
MDL Number MFCD00013392
InChI Key ODVREDGIWSDQFD-UHFFFAOYSA-N
Synonym 1,3-bis tosyloxy propane, propane-1,3-diyl bis 4-methylbenzenesulfonate, 1,3-propanediol di-p-toluenesulfonate, 1,3-propanediol di-p-tosylate, 1,3-propanediol-di-4-tosylate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzene-1-sulfonate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzenesulfonate, propane-1,3-di-tos, acmc-209liv, 1,3-propanediol ditosylate
PubChem CID 230370
IUPAC Name 3-[(4-methylbenzenesulfonyl)oxy]propyl 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCCOS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 92°C
Color White
Quantity 5 g
Formula Weight 384.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(tosyloxy)propane