missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(tosyloxy)-2,2-dimethylpropane 98.0+%, TCI America™

Catalog No. p-7119321
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18705G 5 g
B187025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18705G Supplier TCI America Supplier No. B18705G
Only null left

Chemical Identifiers

CAS 22308-12-9
Molecular Formula C19H24O6S2
Molecular Weight (g/mol) 412.515
MDL Number MFCD00110766
InChI Key LJISGCCLDDOYIJ-UHFFFAOYSA-N
Synonym 1,3-Bis(p-toluenesulfonyloxy)-2,2-dimethylpropane, 2,2-Dimethyl-1,3-propanediol Ditosylate, Neopentyl Glycol Ditosylate
PubChem CID 264253
IUPAC Name [2,2-dimethyl-3-(4-methylphenyl)sulfonyloxypropyl] 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC(C)(C)COS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 121°C
Color White
Quantity 5 g
Formula Weight 412.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(tosyloxy)-2,2-dimethylpropane