missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(aminopropyl)tetramethyldisiloxane, 94%

CAS: 2469-55-8 | C10H28N2OSi2 | 248.52 g/mol

$143.52 - $413.08

Chemical Identifiers

CAS 2469-55-8
Molecular Formula C10H28N2OSi2
Molecular Weight (g/mol) 248.52
MDL Number MFCD00039786
InChI Key GPXCORHXFPYJEH-UHFFFAOYSA-N
Synonym 1,3-bis 3-aminopropyl tetramethyldisiloxane, 3,3'-1,1,3,3-tetramethyldisiloxane-1,3-diyl bis propan-1-amine, 1,3-bis 3-aminopropyl-1,1,3,3-tetramethyldisiloxane, tetramethyl-1,3-bis 3-aminopropyl disiloxane, 1-propanamine, 3,3'-1,1,3,3-tetramethyl-1,3-disiloxanediyl bis, oxybis 3-aminopropyl dimethylsilane, bis 3-aminopropyl tetramethyl disiloxane, silane, oxybis 3-aminopropyl dimethyl, bis gamma-aminopropyl tetramethyldisiloxane, bis 3-aminopropyl tetramethyldisiloxane
PubChem CID 17181
IUPAC Name 3-[[3-aminopropyl(dimethyl)silyl]oxy-dimethylsilyl]propan-1-amine
SMILES C[Si](C)(CCCN)O[Si](C)(C)CCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1729514
View Documents
Thermo Scientific Chemicals
L1729514
25 g
Each for $143.52
Only null left
 
AAL1729522
View Documents
Thermo Scientific Chemicals
L1729522
100 g
Each for $413.08
Only null left
 
Description

Description

1,3-Bis(aminopropyl)tetramethyldisiloxane is used as a curing agent for epoxy molding compounds, which is utilized for high-reliability semiconductor devices. It is also used to prepare siloxane- urea copymers from 4,4’-diphenylmethane diisocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(aminopropyl)tetramethyldisiloxane is used as a curing agent for epoxy molding compounds, which is utilized for high-reliability semiconductor devices. It is also used to prepare siloxane- urea copymers from 4,4’-diphenylmethane diisocyanate.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Incompatible with strong oxidizing agents. Store in a cool place.
Specifications

Chemical Identifiers

2469-55-8
248.52
GPXCORHXFPYJEH-UHFFFAOYSA-N
17181
C[Si](C)(CCCN)O[Si](C)(C)CCCN
C10H28N2OSi2
MFCD00039786
1,3-bis 3-aminopropyl tetramethyldisiloxane, 3,3'-1,1,3,3-tetramethyldisiloxane-1,3-diyl bis propan-1-amine, 1,3-bis 3-aminopropyl-1,1,3,3-tetramethyldisiloxane, tetramethyl-1,3-bis 3-aminopropyl disiloxane, 1-propanamine, 3,3'-1,1,3,3-tetramethyl-1,3-disiloxanediyl bis, oxybis 3-aminopropyl dimethylsilane, bis 3-aminopropyl tetramethyl disiloxane, silane, oxybis 3-aminopropyl dimethyl, bis gamma-aminopropyl tetramethyldisiloxane, bis 3-aminopropyl tetramethyldisiloxane
3-[[3-aminopropyl(dimethyl)silyl]oxy-dimethylsilyl]propan-1-amine

Specifications

132°C to 139°C (11mmHg)
25 g
MFCD00039786
1760876
1,3-bis 3-aminopropyl tetramethyldisiloxane, 3,3'-1,1,3,3-tetramethyldisiloxane-1,3-diyl bis propan-1-amine, 1,3-bis 3-aminopropyl-1,1,3,3-tetramethyldisiloxane, tetramethyl-1,3-bis 3-aminopropyl disiloxane, 1-propanamine, 3,3'-1,1,3,3-tetramethyl-1,3-disiloxanediyl bis, oxybis 3-aminopropyl dimethylsilane, bis 3-aminopropyl tetramethyl disiloxane, silane, oxybis 3-aminopropyl dimethyl, bis gamma-aminopropyl tetramethyldisiloxane, bis 3-aminopropyl tetramethyldisiloxane
GPXCORHXFPYJEH-UHFFFAOYSA-N
3-[[3-aminopropyl(dimethyl)silyl]oxy-dimethylsilyl]propan-1-amine
17181
94%
Air Sensitive
1,3-Bis(aminopropyl)tetramethyldisiloxane
2469-55-8
C10H28N2OSi2
UN2735
91°C (195°F)
Slightly soluble in water.
C[Si](C)(CCCN)O[Si](C)(C)CCCN
248.52
248.52
1.45
0.897
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 219-588-7

RTECSNumber : JM9235000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only