missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(aminopropyl)tetramethyldisiloxane, 94%
Description
1,3-Bis(aminopropyl)tetramethyldisiloxane is used as a curing agent for epoxy molding compounds, which is utilized for high-reliability semiconductor devices. It is also used to prepare siloxane- urea copymers from 4,4’-diphenylmethane diisocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Bis(aminopropyl)tetramethyldisiloxane is used as a curing agent for epoxy molding compounds, which is utilized for high-reliability semiconductor devices. It is also used to prepare siloxane- urea copymers from 4,4’-diphenylmethane diisocyanate.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Incompatible with strong oxidizing agents. Store in a cool place.
1,3-Bis(aminopropyl)tetramethyldisiloxane is used as a curing agent for epoxy molding compounds, which is utilized for high-reliability semiconductor devices. It is also used to prepare siloxane- urea copymers from 4,4’-diphenylmethane diisocyanate.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Incompatible with strong oxidizing agents. Store in a cool place.
Specifications
Specifications
| Boiling Point | 132°C to 139°C (11mmHg) |
| Quantity | 100 g |
| Flash Point | 91°C (195°F) |
| Refractive Index | 1.45 |
| Sensitivity | Air Sensitive |
| Density | 0.897 |
| UN Number | UN2735 |
| Beilstein | 1760876 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 248.52 |
| Show More |
Safety and Handling
| Hazard Category | H227-H314-H335 |
| Hazard Statement | GHS H Statement H314-H227 Causes severe skin burns and eye damage. Combustible liquid. |
| Precautionary Statement | P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
| EINECSNumber | 219-588-7 |
| RTECSNumber | JM9235000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only