missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(aminomethyl)cyclohexane (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. B100525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B100525ML 25 mL
B1005500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B100525ML Supplier TCI America Supplier No. B100525ML

Chemical Identifiers

CAS 2579-20-6
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00001522
InChI Key QLBRROYTTDFLDX-UHFFFAOYSA-N
Synonym 1,3-cyclohexanedimethanamine, 1,3-cyclohexanebis methylamine, 1,3-bis aminomethyl cyclohexane, cyclohexane-1,3-diyldimethanamine, 1,3-di aminomethyl cyclohexane, kodak silver halide solvent hs-103, cyclohexane, 1,3-bis aminomethyl, 3-aminomethyl cyclohexyl methanamine, 3-aminomethyl cyclohexyl methylamine
PubChem CID 17406
IUPAC Name [3-(aminomethyl)cyclohexyl]methanamine
SMILES C1CC(CC(C1)CN)CN

Specifications

Color Colorless
Quantity 25 mL
UN Number 2735
Formula Weight 142.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,3-Bis(aminomethyl)cyclohexane (cis- and trans- mixture)
EINECSNumber (3)-2279
RTECSNumber GU7000000
TSCA Yes