missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-piperidinyl)propane, 97+%

Catalog No. AAL2044222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2044222 100 g
AAL2044214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2044222 Supplier Thermo Scientific Chemicals Supplier No. L2044222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16898-52-5 | C13H26N2 | 210.365 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16898-52-5
Molecular Formula C13H26N2
Molecular Weight (g/mol) 210.365
MDL Number MFCD00038013
InChI Key OXEZLYIDQPBCBB-UHFFFAOYSA-N
Synonym 1,3-bis 4-piperidyl propane, 4,4'-trimethylenedipiperidine, 1,3-di-4-piperidylpropane, piperidine, 4,4'-1,3-propanediyl bis, di-pip, 4-3-piperidin-4-yl propyl piperidine, 4-3-4-piperidyl propyl piperidine, unii-eu53ec219i, 1,3-di 4-piperidyl propane, 1,3-bis 4-piperidinyl propane
PubChem CID 85631
IUPAC Name 4-(3-piperidin-4-ylpropyl)piperidine
SMILES C1CNCCC1CCCC2CCNCC2

Specifications

Melting Point 63°C to 68°C
Boiling Point 327°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 111615
Sensitivity Hygroscopic
Formula Weight 210.36
Percent Purity ≥97%
Chemical Name or Material 1,3-Bis(4-piperidinyl)propane
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 240-941-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only