missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-methoxyphenyl)-1,3-propanedione 98.0+%, TCI America™

Catalog No. B17335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17335G 5 g
B173325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17335G Supplier TCI America Supplier No. B17335G
Only null left

Chemical Identifiers

CAS 18362-51-1
Molecular Formula C17H16O4
Molecular Weight (g/mol) 284.31
MDL Number MFCD00025817
InChI Key GNMDORSUZRRMFS-UHFFFAOYSA-N
PubChem CID 87596
IUPAC Name 1,3-bis(4-methoxyphenyl)propane-1,3-dione
SMILES COC1=CC=C(C=C1)C(=O)CC(=O)C1=CC=C(OC)C=C1

Specifications

Melting Point 116°C
Quantity 5 g
Formula Weight 284.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(4-methoxyphenyl)-1,3-propanedione