missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4,5-dihydro-2-oxazolyl)benzene 98.0+%, TCI America™

Catalog No. p-7119086
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B151125G 25 g
B1511100G 100 g
B1511500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B151125G Supplier TCI America Supplier No. B151125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34052-90-9
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD00191606
InChI Key HMOZDINWBHMBSQ-UHFFFAOYSA-N
PubChem CID 604269
IUPAC Name 2-[3-(4,5-dihydro-1,3-oxazol-2-yl)phenyl]-4,5-dihydro-1,3-oxazole
SMILES C1CN=C(O1)C1=CC(=CC=C1)C1=NCCO1

Specifications

Melting Point 146°C
Quantity 25 g
Synonym 2,2′C-(1,3-Phenylene)bis(2-oxazoline)
Formula Weight 216.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(4,5-dihydro-2-oxazolyl)benzene
EINECSNumber (5)-5683
TSCA Yes