missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(3-aminopropyl)tetramethyldisiloxane 83.0+%, TCI America™

$48.46 - $124.89

Chemical Identifiers

CAS 2469-55-8
Molecular Formula C10H28N2OSi2
Molecular Weight (g/mol) 248.52
MDL Number MFCD00039786
InChI Key GPXCORHXFPYJEH-UHFFFAOYSA-N
Synonym 1,3-bis 3-aminopropyl tetramethyldisiloxane, 3,3'-1,1,3,3-tetramethyldisiloxane-1,3-diyl bis propan-1-amine, 1,3-bis 3-aminopropyl-1,1,3,3-tetramethyldisiloxane, tetramethyl-1,3-bis 3-aminopropyl disiloxane, 1-propanamine, 3,3'-1,1,3,3-tetramethyl-1,3-disiloxanediyl bis, oxybis 3-aminopropyl dimethylsilane, bis 3-aminopropyl tetramethyl disiloxane, silane, oxybis 3-aminopropyl dimethyl, bis gamma-aminopropyl tetramethyldisiloxane, bis 3-aminopropyl tetramethyldisiloxane
PubChem CID 17181
IUPAC Name (3-aminopropyl)({[(3-aminopropyl)dimethylsilyl]oxy})dimethylsilane
SMILES C[Si](C)(CCCN)O[Si](C)(C)CCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B15525G
View Documents
TCI America
B15525G
5 g
Each for $48.46
Only null left
 
B155225G
View Documents
TCI America
B155225G
25 g
Each for $124.89
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2469-55-8
248.52
GPXCORHXFPYJEH-UHFFFAOYSA-N
17181
C[Si](C)(CCCN)O[Si](C)(C)CCCN
C10H28N2OSi2
MFCD00039786
1,3-bis 3-aminopropyl tetramethyldisiloxane, 3,3'-1,1,3,3-tetramethyldisiloxane-1,3-diyl bis propan-1-amine, 1,3-bis 3-aminopropyl-1,1,3,3-tetramethyldisiloxane, tetramethyl-1,3-bis 3-aminopropyl disiloxane, 1-propanamine, 3,3'-1,1,3,3-tetramethyl-1,3-disiloxanediyl bis, oxybis 3-aminopropyl dimethylsilane, bis 3-aminopropyl tetramethyl disiloxane, silane, oxybis 3-aminopropyl dimethyl, bis gamma-aminopropyl tetramethyldisiloxane, bis 3-aminopropyl tetramethyldisiloxane
(3-aminopropyl)({[(3-aminopropyl)dimethylsilyl]oxy})dimethylsilane

Specifications

2469-55-8
142°C
MFCD00039786
2735
GPXCORHXFPYJEH-UHFFFAOYSA-N
(3-aminopropyl)({[(3-aminopropyl)dimethylsilyl]oxy})dimethylsilane
17181
≥83.0% (GC)
1,3-Bis(3-aminopropyl)tetramethyldisiloxane [Monomer for silicon modified polyamides]
Yellow
C10H28N2OSi2
5 g
1,3-bis 3-aminopropyl tetramethyldisiloxane, 3,3'-1,1,3,3-tetramethyldisiloxane-1,3-diyl bis propan-1-amine, 1,3-bis 3-aminopropyl-1,1,3,3-tetramethyldisiloxane, tetramethyl-1,3-bis 3-aminopropyl disiloxane, 1-propanamine, 3,3'-1,1,3,3-tetramethyl-1,3-disiloxanediyl bis, oxybis 3-aminopropyl dimethylsilane, bis 3-aminopropyl tetramethyl disiloxane, silane, oxybis 3-aminopropyl dimethyl, bis gamma-aminopropyl tetramethyldisiloxane, bis 3-aminopropyl tetramethyldisiloxane
C[Si](C)(CCCN)O[Si](C)(C)CCCN
248.52
248.52
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (7)-0453

RTECSNumber : JM9235000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'