missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2,6-diisopropylphenyl)imidazolium Chloride 98.0+%, TCI America™

Catalog No. D3611500MG
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3611500MG 500 mg
D36115G 5 g
D361125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D3611500MG Supplier TCI America Supplier No. D3611500MG

Chemical Identifiers

CAS 250285-32-6
Molecular Formula C27H37ClN2
Molecular Weight (g/mol) 425.06
MDL Number MFCD02684545
InChI Key AVJBQMXODCVJCJ-UHFFFAOYSA-M
Synonym 1,3-bis 2,6-diisopropylphenyl imidazolium chloride, 1,3-bis 2,6-diisopropylphenyl-1h-imidazol-3-ium chloride, 1,3-bis 2,6-di-i-propylphenyl imidazolium chloride, ipr hcl, 2,5-bis 2,6-diisopropylphenyl imidazolium chloride, ipr.hcl, 1,3-bis 2,6-bis 1-methylethyl phenyl-1h-imidazolium chloride, 1,3-bis 2,6-di-i-propylphenyl imidazoliumchloride, 1,3-bis-2,6-diisopropyl-phenyl-3h-imidazol-1-ium chloride, 1,3-bis 2,6-bis prop-2-yl phenyl-1h-imidazol-3-ium chloride
PubChem CID 2734913
IUPAC Name 1,3-bis[2,6-bis(propan-2-yl)phenyl]-3H-1λ⁵-imidazol-1-ylium chloride
SMILES [Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C

Specifications

Melting Point 278°C
Quantity 500 mg
Formula Weight 425.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(2,6-diisopropylphenyl)imidazolium Chloride