missing translation for 'onlineSavingsMsg'
Learn More

1,3-bis(2,6-diisopropylphenyl)imidazolium chloride, 97%

Catalog No. AC378310020
Change view
Click to view available options
Quantity:
2 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378310020 2 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC378310020 Supplier Thermo Scientific Chemicals Supplier No. 378310020
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 250285-32-6 | C27H37ClN2 | 425.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 250285-32-6
Molecular Formula C27H37ClN2
Molecular Weight (g/mol) 425.06
MDL Number MFCD02684545
InChI Key AVJBQMXODCVJCJ-UHFFFAOYSA-M
Synonym 1,3-bis 2,6-diisopropylphenyl imidazolium chloride, 1,3-bis 2,6-diisopropylphenyl-1h-imidazol-3-ium chloride, 1,3-bis 2,6-di-i-propylphenyl imidazolium chloride, ipr hcl, 2,5-bis 2,6-diisopropylphenyl imidazolium chloride, ipr.hcl, 1,3-bis 2,6-bis 1-methylethyl phenyl-1h-imidazolium chloride, 1,3-bis 2,6-di-i-propylphenyl imidazoliumchloride, 1,3-bis-2,6-diisopropyl-phenyl-3h-imidazol-1-ium chloride, 1,3-bis 2,6-bis prop-2-yl phenyl-1h-imidazol-3-ium chloride
PubChem CID 2734913
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]imidazol-1-ium;chloride
SMILES [Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Tan or White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 2 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 425.06
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 3-Bis(2, 6-diisopropylphenyl)imidazolium chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only