missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2,4,6-trimethylphenyl)imidazolinium Chloride 98.0+%, TCI America™

Catalog No. B31585G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31585G 5 g
B31581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31585G Supplier TCI America Supplier No. B31585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 173035-10-4
Molecular Formula C21H27ClN2
Molecular Weight (g/mol) 342.91
MDL Number MFCD09039279
InChI Key COGMCBFILULEOS-UHFFFAOYSA-M
Synonym 1,3-dimesitylimidazolidine hydrochloride, 1,3-bis 2,4,6-trimethylphenyl-imidazolidinium-chloride, 1,3-dimesitylimidazolidine, chloride salt, 1,3-bis-2,4,6-trimethyl-phenyl-imidazolidin-1-ium chloride, 1,3-bis 2,4,6-trimethylphenyl imidazolidine hydrochloride
IUPAC Name 1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydro-3H-1λ⁵-imidazol-1-ylium chloride
SMILES [Cl-].CC1=CC(C)=C(N2CC[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1

Specifications

Melting Point 286°C
Color White-Yellow
Quantity 5 g
Formula Weight 342.91
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(2,4,6-trimethylphenyl)imidazolinium Chloride