Learn More
1,3-bis(2,4,6-trimethylphenyl)imidazolidinium chloride, 97%
CAS: 173035-10-4 | C21H27ClN2 | 342.91 g/mol
$158.78 - $158.78
Chemical Identifiers
| CAS | 173035-10-4 |
|---|---|
| Molecular Formula | C21H27ClN2 |
| Molecular Weight (g/mol) | 342.91 |
| MDL Number | MFCD09039279 |
| InChI Key | COGMCBFILULEOS-UHFFFAOYSA-M |
| Synonym | 1,3-dimesitylimidazolidine hydrochloride, 1,3-bis 2,4,6-trimethylphenyl-imidazolidinium-chloride, 1,3-dimesitylimidazolidine, chloride salt, 1,3-bis-2,4,6-trimethyl-phenyl-imidazolidin-1-ium chloride, 1,3-bis 2,4,6-trimethylphenyl imidazolidine hydrochloride |
| IUPAC Name | 1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydro-3H-1λâµ-imidazol-1-ylium chloride |
| SMILES | [Cl-].CC1=CC(C)=C(N2CC[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC378330010
|
Thermo Scientific Chemicals
378330010 |
1 g | Glass bottle |
Each for $158.78
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 173035-10-4 | |
| 342.91 | |
| COGMCBFILULEOS-UHFFFAOYSA-M | |
| 1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydro-3H-1λâµ-imidazol-1-ylium chloride |
| C21H27ClN2 | |
| MFCD09039279 | |
| 1,3-dimesitylimidazolidine hydrochloride, 1,3-bis 2,4,6-trimethylphenyl-imidazolidinium-chloride, 1,3-dimesitylimidazolidine, chloride salt, 1,3-bis-2,4,6-trimethyl-phenyl-imidazolidin-1-ium chloride, 1,3-bis 2,4,6-trimethylphenyl imidazolidine hydrochloride | |
| [Cl-].CC1=CC(C)=C(N2CC[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1 |
Specifications
| 173035-10-4 | |
| 100.0 | |
| Beige to Brown-Pink | |
| 96% min. (HPLC) | |
| C21H27ClN2 | |
| 1 g | |
| COGMCBFILULEOS-UHFFFAOYSA-M | |
| 1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydro-3H-1λâµ-imidazol-1-ylium chloride | |
| 342.91 | |
| Powder |
| 96.0 | |
| 280°C to 286°C | |
| Authentic | |
| Glass bottle | |
| MFCD09039279 | |
| 1,3-dimesitylimidazolidine hydrochloride, 1,3-bis 2,4,6-trimethylphenyl-imidazolidinium-chloride, 1,3-dimesitylimidazolidine, chloride salt, 1,3-bis-2,4,6-trimethyl-phenyl-imidazolidin-1-ium chloride, 1,3-bis 2,4,6-trimethylphenyl imidazolidine hydrochloride | |
| [Cl-].CC1=CC(C)=C(N2CC[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1 | |
| 342.91 | |
| 97% | |
| 1,3-Bis(2,4,6-trimethylphenyl)imidazolidinium chloride |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
RUO – Research Use Only