missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzothiazol-2-ylmethylamine hydrochloride, ≥97%, Thermo Scientific™

Catalog No. CC06013CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC06013CB 250 mg Amber Glass Bottle
CC06013DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC06013CB Supplier Thermo Scientific Chemicals Supplier No. CC06013CB

Chemical Identifiers

CAS 29198-41-2
Molecular Formula C8H9ClN2S
Molecular Weight (g/mol) 200.684
MDL Number MFCD03783535
InChI Key WCZDQDCFSDCIIC-UHFFFAOYSA-N
Synonym 1,3-benzothiazol-2-ylmethylamine hydrochloride, benzo d thiazol-2-ylmethanamine hydrochloride, 1,3-benzothiazol-2-ylmethanamine hydrochloride, 2-aminomethyl-1,3-benzothiazole hydrochloride, 1-1,3-benzothiazol-2-yl methanamine hydrochloride, 1,3-benzothiazol-2-yl-methanamine hydrochloride, c8h8n2s.clh, bestipharma 508-458, benzothiazol-2-ylmethylamine, chloride, benzothiazol-2-ylmethylamine hcl
PubChem CID 2776261
IUPAC Name 1,3-benzothiazol-2-ylmethanamine;hydrochloride
SMILES C1=CC=C2C(=C1)N=C(S2)CN.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 200.69
Percent Purity 95%
Chemical Name or Material 1, 3-Benzothiazol-2-ylmethylamine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.