missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxole-4-carboxylic acid, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 5768-39-8
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD01076411
InChI Key DBUAYOWCIUQXQW-UHFFFAOYSA-N
Synonym benzo d 1,3 dioxole-4-carboxylic acid, 2h-1,3-benzodioxole-4-carboxylic acid, 1,3-benzodioxole-4-carboxylicacid, 2,3-methylenedioxybenzoic acid, 2,3-methylenedioxy benzoic acid, o-piperonylic acid, 2-methylenedioxybenzoic acid, 4-carboxy-1,3-benzodioxole, 1-carboxy-methylenedioxybenzene, benzo 1,3 dioxole-4-carboxylic acid
PubChem CID 304832
IUPAC Name 1,3-benzodioxole-4-carboxylic acid
SMILES C1OC2=CC=CC(=C2O1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC01701CB
View Documents
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC01701DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

5768-39-8
166.132
DBUAYOWCIUQXQW-UHFFFAOYSA-N
304832
C1OC2=CC=CC(=C2O1)C(=O)O
C8H6O4
MFCD01076411
benzo d 1,3 dioxole-4-carboxylic acid, 2h-1,3-benzodioxole-4-carboxylic acid, 1,3-benzodioxole-4-carboxylicacid, 2,3-methylenedioxybenzoic acid, 2,3-methylenedioxy benzoic acid, o-piperonylic acid, 2-methylenedioxybenzoic acid, 4-carboxy-1,3-benzodioxole, 1-carboxy-methylenedioxybenzene, benzo 1,3 dioxole-4-carboxylic acid
1,3-benzodioxole-4-carboxylic acid

Specifications

5768-39-8
100.0
C8H6O4
benzo d 1,3 dioxole-4-carboxylic acid, 2h-1,3-benzodioxole-4-carboxylic acid, 1,3-benzodioxole-4-carboxylicacid, 2,3-methylenedioxybenzoic acid, 2,3-methylenedioxy benzoic acid, o-piperonylic acid, 2-methylenedioxybenzoic acid, 4-carboxy-1,3-benzodioxole, 1-carboxy-methylenedioxybenzene, benzo 1,3 dioxole-4-carboxylic acid
C1OC2=CC=CC(=C2O1)C(=O)O
166.132
166.13
1, 3-Benzodioxole-4-carboxylic acid
96.5
97%
MFCD01076411
DBUAYOWCIUQXQW-UHFFFAOYSA-N
1,3-benzodioxole-4-carboxylic acid
304832
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents