missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxol-5-ylacetyl chloride, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC50802EA

 View more versions of this product

Catalog No. CC50802EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1, 3-Benzodioxol-5-ylacetyl chloride
96.5
97%
MFCD03424722
benzo 1,3 dioxol-5-yl-acetyl chloride, 1,3-benzodioxol-5-ylacetyl chloride, 2-benzo d 1,3 dioxol-5-yl acetyl chloride, benzo 1,3 dioxol-5-ylacetyl chloride, 2-2h-1,3-benzodioxol-5-yl acetyl chloride, 2-2h-benzo 3,4-d 1,3-dioxolen-5-yl acetyl chloride, 1,3-benzodioxole-5-acetylchloride, 3,4-methylenedioxyphenylacetylchloride, 3,4-methylenedioxyphenylacetyl chloride
ClC(=O)CC1=CC2=C(OCO2)C=C1
198.60
198.6
6845-81-4
100.0
C9H7ClO3
10g
HGSZGCXMAJSUSC-UHFFFAOYSA-N
2-(2H-1,3-benzodioxol-5-yl)acetyl chloride
2756584
97%

Chemical Identifiers

6845-81-4
198.60
HGSZGCXMAJSUSC-UHFFFAOYSA-N
2756584
ClC(=O)CC1=CC2=C(OCO2)C=C1
C9H7ClO3
MFCD03424722
benzo 1,3 dioxol-5-yl-acetyl chloride, 1,3-benzodioxol-5-ylacetyl chloride, 2-benzo d 1,3 dioxol-5-yl acetyl chloride, benzo 1,3 dioxol-5-ylacetyl chloride, 2-2h-1,3-benzodioxol-5-yl acetyl chloride, 2-2h-benzo 3,4-d 1,3-dioxolen-5-yl acetyl chloride, 1,3-benzodioxole-5-acetylchloride, 3,4-methylenedioxyphenylacetylchloride, 3,4-methylenedioxyphenylacetyl chloride
2-(2H-1,3-benzodioxol-5-yl)acetyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions