missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Benzenedisulfonyl Chloride 98.0+%, TCI America™
$95.06 - $285.21
Chemical Identifiers
| CAS | 585-47-7 |
|---|---|
| Molecular Formula | C6H4Cl2O4S2 |
| Molecular Weight (g/mol) | 275.11 |
| MDL Number | MFCD00041879 |
| InChI Key | ALIQZUMMPOYCIS-UHFFFAOYSA-N |
| Synonym | 1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a |
| PubChem CID | 96240 |
| IUPAC Name | benzene-1,3-disulfonyl dichloride |
| SMILES | ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O |
Chemical Identifiers
| 585-47-7 | |
| 275.11 | |
| ALIQZUMMPOYCIS-UHFFFAOYSA-N | |
| 96240 | |
| ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O |
| C6H4Cl2O4S2 | |
| MFCD00041879 | |
| 1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a | |
| benzene-1,3-disulfonyl dichloride |
Specifications
| 585-47-7 | |
| 145°C | |
| MFCD00041879 | |
| 3261 | |
| ALIQZUMMPOYCIS-UHFFFAOYSA-N | |
| benzene-1,3-disulfonyl dichloride | |
| 96240 | |
| ≥98.0% (HPLC,T) | |
| 1,3-Benzenedisulfonyl Chloride |
| 62°C | |
| C6H4Cl2O4S2 | |
| 5 g | |
| 1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a | |
| ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O | |
| 275.11 | |
| 275.11 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1899
TSCA : Yes
Recommended Storage : Refrigerator