missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedisulfonyl chloride, 97%

CAS: 585-47-7 | C6H4Cl2O4S2 | 275.11 g/mol

$137.52 - $1511.10

Chemical Identifiers

CAS 585-47-7
Molecular Formula C6H4Cl2O4S2
Molecular Weight (g/mol) 275.11
MDL Number MFCD00041879
InChI Key ALIQZUMMPOYCIS-UHFFFAOYSA-N
Synonym 1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a
PubChem CID 96240
SMILES ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2404406
View Documents
Thermo Scientific Chemicals
B2404406
5 g
Each for $137.52
Only null left
 
AAB2404414
View Documents
Thermo Scientific Chemicals
B2404414
25 g
Each for $492.52
Only null left
 
AAB2404422
View Documents
Thermo Scientific Chemicals
B2404422
100 g
Each for $1,511.10
Only null left
 
Description

Description

1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

Solubility
Soluble in DMSO and methanol. Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.
Specifications

Chemical Identifiers

585-47-7
275.11
ALIQZUMMPOYCIS-UHFFFAOYSA-N
96240
C6H4Cl2O4S2
MFCD00041879
1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a
ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O

Specifications

585-47-7
C6H4Cl2O4S2
5 g
1118508
1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a
ALIQZUMMPOYCIS-UHFFFAOYSA-N
benzene-1,3-disulfonyl dichloride
96240
98%
58°C to 60°C
MFCD00041879
UN3261
Moisture sensitive
Soluble in DMSO and methanol. Reacts with water.
ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O
275.11
275.13
1,3-Benzenedisulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 209-556-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only