missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedisulfonyl chloride, 97%

Catalog No. AAB2404406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2404406 5 g
AAB2404414 25 g
AAB2404422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2404406 Supplier Thermo Scientific Chemicals Supplier No. B2404406

CAS: 585-47-7 | C6H4Cl2O4S2 | 275.11 g/mol

1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

Solubility
Soluble in DMSO and methanol. Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.

Chemical Identifiers

CAS 585-47-7
Molecular Formula C6H4Cl2O4S2
Molecular Weight (g/mol) 275.11
MDL Number MFCD00041879
InChI Key ALIQZUMMPOYCIS-UHFFFAOYSA-N
Synonym 1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a
PubChem CID 96240
SMILES ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O

Specifications

Melting Point 58°C to 60°C
Quantity 5 g
UN Number UN3261
Beilstein 1118508
Sensitivity Moisture sensitive
Solubility Information Soluble in DMSO and methanol. Reacts with water.
IUPAC Name benzene-1,3-disulfonyl dichloride
Formula Weight 275.13
Percent Purity 98%
Chemical Name or Material 1,3-Benzenedisulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 209-556-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only