missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedimethanol, 98%, Thermo Scientific™

Chemical Identifiers

CAS 626-18-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004649
InChI Key YWMLORGQOFONNT-UHFFFAOYSA-N
Synonym 1,3-benzenedimethanol, 3-hydroxymethyl phenyl methanol, 1,3-phenylenedimethanol, 1,3-dihydroxymethylbenzene, m-xylene-alpha,alpha'-diol, m-xylene glycol, m-xylylene glycol, 3-hydroxymethyl phenyl methan-1-ol, m-xylylenalkohol, m-benzenedimethanol
PubChem CID 69374
IUPAC Name [3-(hydroxymethyl)phenyl]methanol
SMILES C1=CC(=CC(=C1)CO)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC188200020
SDS View Documents
Thermo Scientific Chemicals
188200020
2g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC188200100
SDS View Documents
Thermo Scientific Chemicals
188200100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

626-18-6
138.17
YWMLORGQOFONNT-UHFFFAOYSA-N
69374
C1=CC(=CC(=C1)CO)CO
C8H10O2
MFCD00004649
1,3-benzenedimethanol, 3-hydroxymethyl phenyl methanol, 1,3-phenylenedimethanol, 1,3-dihydroxymethylbenzene, m-xylene-alpha,alpha'-diol, m-xylene glycol, m-xylylene glycol, 3-hydroxymethyl phenyl methan-1-ol, m-xylylenalkohol, m-benzenedimethanol
[3-(hydroxymethyl)phenyl]methanol

Specifications

626-18-6
100.0
Authentic
Glass bottle
C6H4(CH2OH)2
2g
1,3-benzenedimethanol, 3-hydroxymethyl phenyl methanol, 1,3-phenylenedimethanol, 1,3-dihydroxymethylbenzene, m-xylene-alpha,alpha'-diol, m-xylene glycol, m-xylylene glycol, 3-hydroxymethyl phenyl methan-1-ol, m-xylylenalkohol, m-benzenedimethanol
C1=CC(=CC(=C1)CO)CO
138.17
138.17
1, 3-Benzenedimethanol, 98%
97.5
154.0°C to 159.0°C (13.0mmHg)
97.5% min. (GC)
C8H10O2
MFCD00004649
06, 914
YWMLORGQOFONNT-UHFFFAOYSA-N
[3-(hydroxymethyl)phenyl]methanol
69374
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 210-934-2

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only