missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Aminopropyl)pyrrolidine 98.0+%, TCI America™

Catalog No. A22705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22705G 5 g
A227025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22705G Supplier TCI America Supplier No. A22705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23159-07-1
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00014100
InChI Key VPBWZBGZWHDNKL-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-yl propan-1-amine, 1-3-aminopropyl pyrrolidine, 1-3-aminopropyl-pyrrolidine, 1-pyrrolidinepropanamine, n-3-aminopropyl pyrrolidine, 3-pyrrolidinopropylamine, 3-1-pyrrolidinyl propylamine, pyrrolidine, 1-3-aminopropyl, 3-pyrrolidin-1-ylpropylamine, 3-1-pyrrolidino propylamine
PubChem CID 31670
IUPAC Name 3-pyrrolidin-1-ylpropan-1-amine
SMILES C1CCN(C1)CCCN

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
UN Number 2735
Formula Weight 128.22
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-(3-Aminopropyl)pyrrolidine