missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Adamantanedicarboxylic Acid 97.0+%, TCI America™
$167.76 - $564.81
Chemical Identifiers
| CAS | 39269-10-8 |
|---|---|
| Molecular Formula | C12H14O4 |
| Molecular Weight (g/mol) | 222.24 |
| MDL Number | MFCD00167790 |
| InChI Key | PAVQGHWQOQZQEH-UHFFFAOYSA-L |
| Synonym | 1,3-adamantanedicarboxylic acid, 1,3-dicarboxyadamantane, 1s,3s,5r,7r-adamantane-1,3-dicarboxylic acid, 1,3-adamantane dicarboxylic acid, tricyclo 3.3.1.1 sup 3,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.13,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1,3-dicarboxylic acid, 1r,3s,5s,7s-adamantane-1,3-dicarboxylic acid, 1,3-adamantanedicarboxylicacid |
| PubChem CID | 64339 |
| IUPAC Name | adamantane-1,3-dicarboxylate |
| SMILES | [O-]C(=O)C12CC3CC(C1)CC(C3)(C2)C([O-])=O |
Chemical Identifiers
| 39269-10-8 | |
| 222.24 | |
| PAVQGHWQOQZQEH-UHFFFAOYSA-L | |
| 64339 | |
| [O-]C(=O)C12CC3CC(C1)CC(C3)(C2)C([O-])=O |
| C12H14O4 | |
| MFCD00167790 | |
| 1,3-adamantanedicarboxylic acid, 1,3-dicarboxyadamantane, 1s,3s,5r,7r-adamantane-1,3-dicarboxylic acid, 1,3-adamantane dicarboxylic acid, tricyclo 3.3.1.1 sup 3,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.13,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1,3-dicarboxylic acid, 1r,3s,5s,7s-adamantane-1,3-dicarboxylic acid, 1,3-adamantanedicarboxylicacid | |
| adamantane-1,3-dicarboxylate |
Specifications
| 39269-10-8 | |
| C12H14O4 | |
| 5 g | |
| PAVQGHWQOQZQEH-UHFFFAOYSA-L | |
| adamantane-1,3-dicarboxylate | |
| 64339 | |
| ≥97.0% (GC,T) | |
| 1,3-Adamantanedicarboxylic Acid |
| White-Yellow | |
| MFCD00167790 | |
| 1,3-adamantanedicarboxylic acid, 1,3-dicarboxyadamantane, 1s,3s,5r,7r-adamantane-1,3-dicarboxylic acid, 1,3-adamantane dicarboxylic acid, tricyclo 3.3.1.1 sup 3,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.13,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1,3-dicarboxylic acid, 1r,3s,5s,7s-adamantane-1,3-dicarboxylic acid, 1,3-adamantanedicarboxylicacid | |
| [O-]C(=O)C12CC3CC(C1)CC(C3)(C2)C([O-])=O | |
| 222.24 | |
| 224.26 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : YD1994000
TSCA : No