missing translation for 'onlineSavingsMsg'
Learn More

1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate, mixture of isomers, Thermo Scientific™

Chemical Identifiers

CAS 19437-42-4
Molecular Formula xNa·xH2O
Molecular Weight (g/mol) 368.36
MDL Number MFCD00149263
InChI Key BERQWQSQOIEUMA-UHFFFAOYSA-M
Synonym 1,3,6-naphthalenetrisulfonic acid, sodium salt, 1,3,6-naphthalenetrisulfonic acid, sodium salt 1:?
PubChem CID 73555608
IUPAC Name sodium;5,7-disulfonaphthalene-2-sulfonate
SMILES C1=CC2=C(C=C(C=C2C=C1S(=O)(=O)[O-])S(=O)(=O)O)S(=O)(=O)O.[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC269010010
SDS View Documents
Thermo Scientific Chemicals
269010010
1kg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

19437-42-4
368.36
BERQWQSQOIEUMA-UHFFFAOYSA-M
73555608
C1=CC2=C(C=C(C=C2C=C1S(=O)(=O)[O-])S(=O)(=O)O)S(=O)(=O)O.[Na+]
xNa·xH2O
MFCD00149263
1,3,6-naphthalenetrisulfonic acid, sodium salt, 1,3,6-naphthalenetrisulfonic acid, sodium salt 1:?
sodium;5,7-disulfonaphthalene-2-sulfonate

Specifications

19437-42-4,19437-42-4
Glass bottle
MFCD00149263
11,229
Solubility in water: 600g/L (20°C)
C1=CC2=C(C=C(C=C2C=C1S(=O)(=O)[O-])S(=O)(=O)O)S(=O)(=O)O.[Na+]
368.36
368.36
1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate
8% max.
xNa·xH2O
1kg
1,3,6-naphthalenetrisulfonic acid, sodium salt, 1,3,6-naphthalenetrisulfonic acid, sodium salt 1:?
BERQWQSQOIEUMA-UHFFFAOYSA-M
sodium;5,7-disulfonaphthalene-2-sulfonate
73555608
≥90% (by difference)
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 243-068-9

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only