missing translation for 'onlineSavingsMsg'
Learn More

1,3,6,8-Tetrabromopyrene, 98%

$73.42 - $790.86

Specifications

Melting Point 410°C to 413°C
Percent Purity 98%
Physical Form Solid
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3605303
View Documents
Thermo Scientific Chemicals
H3605303
1 g
Each for $73.42
Only null left
 
AAH3605306
View Documents
Thermo Scientific Chemicals
H3605306
5 g
Each for $211.84
Only null left
 
AAH3605314
View Documents
Thermo Scientific Chemicals
H3605314
25 g
Each for $790.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

410°C to 413°C
98%
Solid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'