missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,6,8-Tetra-tert-butylcarbazole 98.0+%, TCI America™
Supplier: TCI America T3051200MG
Specifications
| 1,3,6,8-Tetra-tert-butylcarbazole | |
| 191°C | |
| C28H41N | |
| 200 mg | |
| CC(C)(C)C1=CC(=C2C(=C1)C3=CC(=CC(=C3N2)C(C)(C)C)C(C)(C)C)C(C)(C)C | |
| 391.643 | |
| 391.64 | |
| Crystalline Powder |
| 34601-54-2 | |
| White-Yellow | |
| MFCD00022212 | |
| OVSGNPWPCZRNKI-UHFFFAOYSA-N | |
| 1,3,6,8-tetratert-butyl-9H-carbazole | |
| 545459 | |
| ≥98.0% (N) |
Chemical Identifiers
| 34601-54-2 | |
| 391.643 | |
| OVSGNPWPCZRNKI-UHFFFAOYSA-N | |
| 1,3,6,8-tetratert-butyl-9H-carbazole |
| C28H41N | |
| MFCD00022212 | |
| 545459 | |
| CC(C)(C)C1=CC(=C2C(=C1)C3=CC(=CC(=C3N2)C(C)(C)C)C(C)(C)C)C(C)(C)C |
Safety and Handling
TSCA : No