missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Tris[4-(diphenylamino)phenyl]benzene 95.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 200 mg |
| Formula Weight | 808.04 |
| Percent Purity | ≥95.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,3,5-Tris[4-(diphenylamino)phenyl]benzene |
Safety and Handling
| TSCA | No |