missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(4-carboxyphenyl)benzene 98.0+%, TCI America™

Catalog No. T26471G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26471G 1 g
T26475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26471G Supplier TCI America Supplier No. T26471G

Chemical Identifiers

CAS 50446-44-1
Molecular Formula C27H18O6
Molecular Weight (g/mol) 438.44
MDL Number MFCD10000888
InChI Key SATWKVZGMWCXOJ-UHFFFAOYSA-N
Synonym 1,3,5-tris 4-carboxyphenyl benzene, 1,3,5-tri 4-carboxyphenyl benzene, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylic acid, 1,1':3',1-terphenyl-4,4-dicarboxylic acid, 5'-4-carboxyphenyl, acmc-209kmn, h3btb, 4,4?,4??,-benzene-1,3,5-triyl-tris benzoic acid, 1,3,5-tris 4-carboxyphenyl benzene, ≤20 wt. % solvent, 3',5'-bis 4-carboxyphenyl-1,1'-biphenyl-4-carboxylic acid, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylicacid
PubChem CID 10694305
IUPAC Name 3',5'-bis(4-carboxyphenyl)-[1,1'-biphenyl]-4-carboxylic acid
SMILES OC(=O)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)C(O)=O)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 325°C
Color White
Quantity 1 g
Formula Weight 438.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(4-carboxyphenyl)benzene