missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris[4-(9-carbazolyl)phenyl]benzene 98.0+%, TCI America™

Catalog No. t3439200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3439200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3439200MG Supplier TCI America Supplier No. T3439200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 160780-82-5
Color White-Yellow
Molecular Formula C60H39N3
Quantity 200 mg
InChI Key XCICDYGIJBPNPC-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)C5=CC(=CC(=C5)C6=CC=C(C=C6)N7C8=CC=CC=C8C9=CC=CC=C97)C1=CC=C(C=C1)N1C2=CC=CC=C2C2=CC=CC=C21
IUPAC Name 9-[4-[3,5-bis(4-carbazol-9-ylphenyl)phenyl]phenyl]carbazole
Molecular Weight (g/mol) 801.993
PubChem CID 18991218
Formula Weight 801.99
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris[4-(9-carbazolyl)phenyl]benzene
Show More Show Less