missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(1-phenyl-1H-benzimidazol-2-yl)benzene 98.0+%, TCI America™

Catalog No. T35371G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35371G 1 g
T3537200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35371G Supplier TCI America Supplier No. T35371G

Chemical Identifiers

CAS 192198-85-9
Molecular Formula C45H30N6
Molecular Weight (g/mol) 654.78
MDL Number MFCD08276433
InChI Key GEQBRULPNIVQPP-UHFFFAOYSA-N
Synonym 2,2′,2′′-(1,3,5-Benzenetriyl)tris(1-phenyl-1H-benzimidazole), TPBi
PubChem CID 21932919
IUPAC Name 2-[3,5-bis(1-phenyl-1H-1,3-benzodiazol-2-yl)phenyl]-1-phenyl-1H-1,3-benzodiazole
SMILES C1=CC=C(C=C1)N1C2=CC=CC=C2N=C1C1=CC(=CC(=C1)C1=NC2=CC=CC=C2N1C1=CC=CC=C1)C1=NC2=CC=CC=C2N1C1=CC=CC=C1

Specifications

Melting Point 275°C
Color White
Quantity 1 g
Formula Weight 654.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(1-phenyl-1H-benzimidazol-2-yl)benzene