missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Triphenylformazan 92.0+%, TCI America™
Supplier: TCI America T017525G
Specifications
| 1,3,5-Triphenylformazan | |
| C19H16N4 | |
| 25 g | |
| BEIHVSJTPTXQGB-QUHCWSQZSA-N | |
| N'-anilino-N-phenyliminobenzenecarboximidamide | |
| 5464164 | |
| ≥92.0% (HPLC) |
| 531-52-2 | |
| MFCD00020647 | |
| Tetrazole Red Formazan, Tetrazolium Red Formazan, TTC Formazan | |
| C1=CC=C(C=C1)C(=NNC2=CC=CC=C2)N=NC3=CC=CC=C3 | |
| 300.365 | |
| 300.37 | |
| Crystalline Powder |
Chemical Identifiers
| 531-52-2 | |
| 300.365 | |
| BEIHVSJTPTXQGB-QUHCWSQZSA-N | |
| 5464164 | |
| C1=CC=C(C=C1)C(=NNC2=CC=CC=C2)N=NC3=CC=CC=C3 |
| C19H16N4 | |
| MFCD00020647 | |
| Tetrazole Red Formazan, Tetrazolium Red Formazan, TTC Formazan | |
| N'-anilino-N-phenyliminobenzenecarboximidamide |
Safety and Handling
TSCA : Yes