missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Trioxane 99.0+%, TCI America™
Supplier: TCI America T0505500G
Specifications
| 1,3,5-Trioxane | |
| 63°C | |
| 115°C | |
| MFCD00006563 | |
| 1325 | |
| BGJSXRVXTHVRSN-UHFFFAOYSA-N | |
| 1,3,5-trioxane | |
| 8081 | |
| 90.08 | |
| Crystalline Powder |
| 110-88-3 | |
| White | |
| C3H6O3 | |
| 500 g | |
| s-trioxane, trioxymethylene, metaformaldehyde, triformol, formaldehyde, trimer, aldeform, formagene, marvosan, trioxan, sym-trioxane | |
| C1OCOCO1 | |
| 90.078 | |
| CHEBI:38043 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 110-88-3 | |
| 90.078 | |
| BGJSXRVXTHVRSN-UHFFFAOYSA-N | |
| 8081 | |
| 1,3,5-trioxane |
| C3H6O3 | |
| MFCD00006563 | |
| s-trioxane, trioxymethylene, metaformaldehyde, triformol, formaldehyde, trimer, aldeform, formagene, marvosan, trioxan, sym-trioxane | |
| CHEBI:38043 | |
| C1OCOCO1 |
Safety and Handling
EINECSNumber : (5)-1084
RTECSNumber : YK0350000
TSCA : Yes