missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde, 98%

Catalog No. AAH5022503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5022503 1 g
AAH5022506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5022503 Supplier Thermo Scientific Chemicals Supplier No. H5022503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2644-93-1 | C7H10N2O | 138.17 g/mol

1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde is used as organic chemical synthesis intermediate.

Notes
Air sensitive. Store away from oxidizing agents.

Chemical Identifiers

CAS 2644-93-1
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00159623
InChI Key HMTUBXVXHHITGO-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde, 1,3,5-trimethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1,3,5-trimethyl, n-methyl-3,5-dimethyl-4-formylpyrazole, 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde?, akos pao-0265, timtec-bb sbb000012, acmc-1cq3n, art-chem-bb b006543, ksc496q8h
PubChem CID 736515
IUPAC Name 1,3,5-trimethylpyrazole-4-carbaldehyde
SMILES CC1=C(C(=NN1C)C)C=O

Specifications

Melting Point 80°C to 83°C
Boiling Point 132°C (12 mmHg)
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material 1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only