missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 138.17 |
| Percent Purity | 97% |
| Chemical Name or Material | 1, 3, 5-Trimethyl-1H-pyrazole-4-carbaldehyde |