missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazol-4-amine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 28466-21-9
Molecular Formula C6H11N3
Molecular Weight (g/mol) 125.175
MDL Number MFCD00052883
InChI Key SSDGMKHZMNTWLS-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazol-4-amine, 4-amino-1,3,5-trimethylpyrazole, trimethyl-1h-pyrazol-4-amine, pyrazole, 4-amino-1,3,5-trimethyl, 4-amino-1,3,5-trimethyl-1h-pyrazole, trimethylpyrazol-4-amine, 1,3,5-trimethyl-1h-pyrazol-4-ylamine, 1h-pyrazol-4-amine, 1,3,5-trimethyl, 1,3,5-trimethyl-1h-pyrazole-4-amine, 1,3,5-trimethylpyrazole-4-ylamine
PubChem CID 161603
IUPAC Name 1,3,5-trimethylpyrazol-4-amine
SMILES CC1=C(C(=NN1C)C)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CD11242DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

28466-21-9
125.175
SSDGMKHZMNTWLS-UHFFFAOYSA-N
161603
CC1=C(C(=NN1C)C)N
C6H11N3
MFCD00052883
1,3,5-trimethyl-1h-pyrazol-4-amine, 4-amino-1,3,5-trimethylpyrazole, trimethyl-1h-pyrazol-4-amine, pyrazole, 4-amino-1,3,5-trimethyl, 4-amino-1,3,5-trimethyl-1h-pyrazole, trimethylpyrazol-4-amine, 1,3,5-trimethyl-1h-pyrazol-4-ylamine, 1h-pyrazol-4-amine, 1,3,5-trimethyl, 1,3,5-trimethyl-1h-pyrazole-4-amine, 1,3,5-trimethylpyrazole-4-ylamine
1,3,5-trimethylpyrazol-4-amine

Specifications

28466-21-9
100.0
C6H11N3
1,3,5-trimethyl-1h-pyrazol-4-amine, 4-amino-1,3,5-trimethylpyrazole, trimethyl-1h-pyrazol-4-amine, pyrazole, 4-amino-1,3,5-trimethyl, 4-amino-1,3,5-trimethyl-1h-pyrazole, trimethylpyrazol-4-amine, 1,3,5-trimethyl-1h-pyrazol-4-ylamine, 1h-pyrazol-4-amine, 1,3,5-trimethyl, 1,3,5-trimethyl-1h-pyrazole-4-amine, 1,3,5-trimethylpyrazole-4-ylamine
CC1=C(C(=NN1C)C)N
125.175
125.17
1, 3, 5-Trimethyl-1H-pyrazol-4-amine
96.5
95%
MFCD00052883
SSDGMKHZMNTWLS-UHFFFAOYSA-N
1,3,5-trimethylpyrazol-4-amine
161603
97%
Videos
SDS
Documents

Documents