missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane, 97%

CAS: 2374-14-3 | C12H21F9O3Si3 | 468.538 g/mol

Supplier:  Thermo Scientific Chemicals L1668014

 View more versions of this product

Catalog No. AAL1668014


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

135°C (22mmHg)
34°C to 36°C
25 g
MFCD00054667
1809096
URZHQOCYXDNFGN-UHFFFAOYSA-N
2,4,6-trimethyl-2,4,6-tris(3,3,3-trifluoropropyl)-1,3,5,2,4,6-trioxatrisilinane
75410
97%
1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane
2374-14-3
C12H21F9O3Si3
UN2811
silicone fluid, fluorosilicone trimer, 1,3,5-trimethyl-1,3,5-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 1,3,5-tris 3,3,3-trifluoropropyl methyl cyclotrisiloxane, cyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl, fluorosilicone cyclic trimer, b071 cut ii, 3,3,3-trifluoropropyl methylcyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 3,3,3-trifluoropropyl methylsiloxane cyclic trimer
C[Si]1(O[Si](O[Si](O1)(C)CCC(F)(F)F)(C)CCC(F)(F)F)CCC(F)(F)F
468.538
468.54
1.24

Chemical Identifiers

2374-14-3
468.538
URZHQOCYXDNFGN-UHFFFAOYSA-N
75410
C[Si]1(O[Si](O[Si](O1)(C)CCC(F)(F)F)(C)CCC(F)(F)F)CCC(F)(F)F
C12H21F9O3Si3
MFCD00054667
silicone fluid, fluorosilicone trimer, 1,3,5-trimethyl-1,3,5-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 1,3,5-tris 3,3,3-trifluoropropyl methyl cyclotrisiloxane, cyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl, fluorosilicone cyclic trimer, b071 cut ii, 3,3,3-trifluoropropyl methylcyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 3,3,3-trifluoropropyl methylsiloxane cyclic trimer
2,4,6-trimethyl-2,4,6-tris(3,3,3-trifluoropropyl)-1,3,5,2,4,6-trioxatrisilinane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P260-P264b-P270-P281-P308+P313-P501c

H361-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 219-154-7

RTECSNumber : GZ4818000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only