missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane, 97%

Catalog No. AAL1668006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1668006 5 g
AAL1668014 25 g
AAL1668022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1668006 Supplier Thermo Scientific Chemicals Supplier No. L1668006

CAS: 2374-14-3 | C12H21F9O3Si3 | 468.538 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2374-14-3
Molecular Formula C12H21F9O3Si3
Molecular Weight (g/mol) 468.538
MDL Number MFCD00054667
InChI Key URZHQOCYXDNFGN-UHFFFAOYSA-N
Synonym silicone fluid, fluorosilicone trimer, 1,3,5-trimethyl-1,3,5-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 1,3,5-tris 3,3,3-trifluoropropyl methyl cyclotrisiloxane, cyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl, fluorosilicone cyclic trimer, b071 cut ii, 3,3,3-trifluoropropyl methylcyclotrisiloxane, 2,4,6-trimethyl-2,4,6-tris 3,3,3-trifluoropropyl cyclotrisiloxane, 3,3,3-trifluoropropyl methylsiloxane cyclic trimer
PubChem CID 75410
IUPAC Name 2,4,6-trimethyl-2,4,6-tris(3,3,3-trifluoropropyl)-1,3,5,2,4,6-trioxatrisilinane
SMILES C[Si]1(O[Si](O[Si](O1)(C)CCC(F)(F)F)(C)CCC(F)(F)F)CCC(F)(F)F

Specifications

Boiling Point 135°C (22mmHg)
Melting Point 34°C to 36°C
Quantity 5 g
UN Number UN2811
Beilstein 1809096
Formula Weight 468.54
Percent Purity 97%
Density 1.24
Chemical Name or Material 1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane
Hazard Category H361-H372
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 219-154-7
RTECSNumber GZ4818000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only