missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triethylbenzene 99.0+%, TCI America™

Catalog No. p-7136469
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08681ML 1 mL
T08685ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08681ML Supplier TCI America Supplier No. T08681ML
Only null left

Chemical Identifiers

CAS 102-25-0
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00009261
InChI Key WJYMPXJVHNDZHD-UHFFFAOYSA-N
Synonym benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade
PubChem CID 7602
IUPAC Name 1,3,5-triethylbenzene
SMILES CCC1=CC(CC)=CC(CC)=C1

Specifications

Color Yellow
Boiling Point 218°C
Quantity 1 mL
UN Number 3082
Formula Weight 162.28
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Triethylbenzene
EINECSNumber (3)-3427
TSCA Yes