missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Triethylbenzene, 95%
Description
,3,5-Triethylbenzene was used as a supramoelcular template to organize molecular-recognition elements. It was also used to synthesize a series of di- and trinucleating ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
,3,5-Triethylbenzene was used as a supramoelcular template to organize molecular-recognition elements. It was also used to synthesize a series of di- and trinucleating ligands.
Solubility
Insoluble in water. Soluble in alcohol, ether.
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
,3,5-Triethylbenzene was used as a supramoelcular template to organize molecular-recognition elements. It was also used to synthesize a series of di- and trinucleating ligands.
Solubility
Insoluble in water. Soluble in alcohol, ether.
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifications
Specifications
| Density | 0.862 |
| Boiling Point | 215°C |
| Flash Point | 76°C (169°F) |
| Refractive Index | 1.495 |
| Quantity | 5 g |
| UN Number | UN3082 |
| Beilstein | 2039481 |
| Solubility Information | Insoluble in water. Soluble in alcohol,ether. |
| Formula Weight | 162.27 |
| Percent Purity | 95% |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H319 |
| Hazard Statement | GHS H Statement H315-H319-H227 Causes skin irritation. Causes serious eye irritation. Combustible liquid. |
| Precautionary Statement | P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S. |
| EINECSNumber | 203-017-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only