missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triazine 98.0+%, TCI America™

Supplier:  TCI America T033925G

 View more versions of this product

Catalog No. T033925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,3,5-Triazine
81°C
114°C
MFCD00006044
s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t
C1=NC=NC=N1
81.08
CHEBI:30259
≥98.0% (GC)
290-87-9
White-Yellow
C3H3N3
25 g
JIHQDMXYYFUGFV-UHFFFAOYSA-N
1,3,5-triazine
9262
81.08
Crystalline Powder

Chemical Identifiers

290-87-9
81.08
JIHQDMXYYFUGFV-UHFFFAOYSA-N
9262
1,3,5-triazine
C3H3N3
MFCD00006044
s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t
CHEBI:30259
C1=NC=NC=N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : XY2957000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions