missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triazine 98.0+%, TCI America™

Catalog No. T03395G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03395G 5 g
T03391G 1 g
T033925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T03395G Supplier TCI America Supplier No. T03395G

Chemical Identifiers

CAS 290-87-9
Molecular Formula C3H3N3
Molecular Weight (g/mol) 81.08
MDL Number MFCD00006044
InChI Key JIHQDMXYYFUGFV-UHFFFAOYSA-N
Synonym s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t
PubChem CID 9262
ChEBI CHEBI:30259
IUPAC Name 1,3,5-triazine
SMILES C1=NC=NC=N1

Specifications

Melting Point 81°C
Color White-Yellow
Boiling Point 114°C
Quantity 5 g
Formula Weight 81.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Triazine
RTECSNumber XY2957000
TSCA Yes