missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triacryloylhexahydro-1,3,5-triazine 98.0+%, TCI America™

Catalog No. p-7136545
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T100025G 25 g
T1000100G 100 g
T1000500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T100025G Supplier TCI America Supplier No. T100025G

Chemical Identifiers

CAS 959-52-4
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
MDL Number MFCD00035738
InChI Key FYBFGAFWCBMEDG-UHFFFAOYSA-N
PubChem CID 70397
IUPAC Name 1-[3,5-di(prop-2-enoyl)-1,3,5-triazinan-1-yl]prop-2-en-1-one
SMILES C=CC(=O)N1CN(CN(C1)C(=O)C=C)C(=O)C=C

Specifications

Melting Point 156°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 249.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Triacryloylhexahydro-1,3,5-triazine
EINECSNumber (5)-1001
RTECSNumber XY9260000
TSCA Yes