missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triacetylbenzene 98.0+%, TCI America™

Catalog No. p-7136721
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13375G 5 g
T133725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13375G Supplier TCI America Supplier No. T13375G

Chemical Identifiers

CAS 779-90-8
Molecular Formula C12H12O3
Molecular Weight (g/mol) 204.225
MDL Number MFCD00008741
InChI Key HSOAIPRTHLEQFI-UHFFFAOYSA-N
Synonym 1,3,5-triacetylbenzene, 1,1',1-benzene-1,3,5-triyl triethanone, 1,3,5,triacetylbenzene, benzene, 1,3,5-triacetyl, 1-3,5-diacetylphenyl ethanone, 1,5-triacetylbenzene, benzene,3,5-triacetyl, acmc-1bc4n, cambridge id 6741393, ethanone, 1,1',1-1,3,5-benzenetriyl tris
PubChem CID 69904
IUPAC Name 1-(3,5-diacetylphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1)C(=O)C)C(=O)C

Specifications

Melting Point 163°C
Color Yellow
Quantity 5 g
Formula Weight 204.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Triacetylbenzene